N-acetylornithine 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
N-Ac-Orn-OH, (S)-2-Acetamido-5-aminopentanoic acid, Ac-Orn-OH, N-Acetyl-L-ornithine
H66648-03
H66648-06
H66648-MD
AAH66648-03EA
93.1
USD
AAH66648-03
AAH66648-06
AAH66648-MD
N-acetylornithine 95%
N-Acetylornithine
N-Acetyl-L-ornithine, 95%
Purity: 95%
CAS No.: 6205-08-9
Molecular Formula: C7H14N2O3
Formula Weight: 174.20
Storage Temperature: -30°C to -10°C
Physical Form: Solid
MDL No.: MFCD00065115
Formula:
C₇H₁₄N₂O₃ Storage Temperature: Refrigerator |
MDL Number:
MFCD00065115 CAS Number: 6205-08-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...