Tetramethyl orthocarbonate 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Tetramethylorthocarbonate
H66617-03
H66617-06
AAH66617-03EA
44.1
USD
AAH66617-03
AAH66617-06
Tetramethyl orthocarbonate 95%
Tetramethyl orthocarbonate
Tetramethyl orthocarbonate, 95%
Purity: 95%
CAS No.: 1850-14-2
Molecular Formula: C5H12O4
Formula Weight: 136.15
Flash Point: 22°(72°F)
Physical Form: Liquid
Density: 1.017
Refractive Index: 1.3850
Sensitivity: Moisture Sensitive
Appearance: Colorless
Melting Point: -5°
Boiling Point: 114°
Beilstein Reference: 1737682
UN No.: 3272
MDL No.: MFCD00008473
Formula:
C₅H₁₂O₄ MW: 136.15 g/mol Boiling Pt: 114 °C Melting Pt: –5 °C Density: 1.017 Flash Pt: 22 °C(72 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008473 CAS Number: 1850-14-2 EINECS: 217-438-5 UN: 3272 ADR: "3",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...