N-Boc-D-aspartic acid-1-methyl ester 97%
Supplier: Thermo Scientific Chemicals
H66573-06
H66573-14
AAH66573-06EA
88.5
USD
AAH66573-06
AAH66573-14
N-Boc-D-aspartic acid-1-methyl ester 97%
N-Boc-D-aspartic acid-1-methyl ester
N-Boc-D-aspartic acid 1-methyl ester, 97%
Purity: 97%
CAS No.: 137130-65-5
Molecular Formula: C10H17NO6
Formula Weight: 247.25
Physical Form: Powder
Appearance: White
Formula:
C₁₀H₁₇NO₆ Storage Temperature: Ambient |
MDL Number:
MFCD03094776 CAS Number: 137130-65-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...