N-(tert-Butoxycarbonyl)-L-methionine N-Succinimidyl Ester 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
Boc-Met-OSu, N-Boc-L-methionine N-Succinimidyl Ester
H66563-06
H66563-14
AAH66563-06EA
53.2
USD
AAH66563-06
AAH66563-14
N-(tert-Butoxycarbonyl)-L-methionine N-Succinimidyl Ester 97%
N-(tert-Butoxycarbonyl)-L-methionine N-succinimidyl ester
N-Boc-L-methionine N-succinimidyl ester, 97%
Purity: 97%
CAS No.: 3845-64-5
Molecular Formula: C14H22N2O6S
Formula Weight: 346.40
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Sensitivity: Moisture Sensitive
Melting Point: 120-128°
Beilstein Reference: 4238574
Optical Rotation: -21°(c=2 in dioxane)
MDL No.: MFCD00042760
Formula:
C₁₄H₂₂N₂O₆S MW: 346.40 g/mol Melting Pt: 120…128 °C Storage Temperature: Freezer |
MDL Number:
MFCD00042760 CAS Number: 3845-64-5 EINECS: 223-341-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...