Fmoc-O-trityl-D-serine 95%

Supplier: Thermo Scientific Chemicals

Synonyms: N-Fmoc-O-trityl-D-serine, Fmoc-D-Ser(Trt)-OH

H66515-06 H66515-14
AAH66515-06EA 153 USD
AAH66515-06 AAH66515-14
Fmoc-O-trityl-D-serine 95%
Fmoc-O-trityl-D-serine

N-Fmoc-O-trityl-D-serine, 95%


Purity: 95% CAS No.: 212688-51-2 Molecular Formula: C37H31NO5 Formula Weight: 569.66 Storage Temperature: -30°C to -10°C Physical Form: Solid MDL No.: MFCD00270546

Formula: C₃₇H₃₁NO₅
MW: 569.657 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00270546
CAS Number: 212688-51-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR