Fmoc-Glu-OAll 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
N-Fmoc-L-glutamic acid 1-allyl ester, Fmoc-Glu-allyl ester, Fmoc-Glu-OAll, 1-Allyl N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-glutamate, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-glutamic acid 1-Allyl ester, 1-Allyl N-Fmoc-L-glutamate, N-Fmoc-L-glutamic acid 1-Allyl ester
N-Fmoc-L-glutamic acid 1-allyl ester, 98%
Purity: 98%
CAS No.: 144120-54-7
Molecular Formula: C23H23NO6
Formula Weight: 409.44
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: Pale brown
Melting Point: 118-122°
Beilstein Reference: 5460531
Optical Rotation: -20°(c=1 in DMF)
MDL No.: MFCD00467718
Formula:
C₂₃H₂₃NO₆ Melting Pt: 118…122 °C Storage Temperature: Ambient |
MDL Number:
MFCD00467718 CAS Number: 144120-54-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...