2-Bromo-N-Fmoc-L-phenylalanine 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
Fmoc-Phe(2-Br)-OH
H66424-06
H66424-14
AAH66424-03EA
28.12
USD
AAH66424-03
AAH66424-06
AAH66424-14
2-Bromo-N-Fmoc-L-phenylalanine 95%
2-Bromo-N-fmoc-L-phenylalanine
2-Bromo-N-Fmoc-L-phenylalanine, 95%
Purity: 95%
CAS No.: 220497-47-2
Molecular Formula: C24H20BrNO4
Formula Weight: 426.47
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: Off-white
MDL No.: MFCD01311743
Formula:
C₂₄H₂₀BrNO₄ MW: 466.331 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD01311743 CAS Number: 220497-47-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...