Fmoc-Asp-OAll 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
N-Fmoc-L-aspartic acid 1-Allyl ester, Fmoc-Asp-allyl ester, N-Fmoc-L-aspartic acid 1-allyl ester, 1-Allyl N-Fmoc-L-aspartate, Fmoc-Asp-OAll, 1-Allyl N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-aspartate, N-[(9H-Fluoren-9-ylmethyloxycarbonyl)]-L-aspartic acid 1-Allyl ester
N-Fmoc-L-aspartic acid 1-allyl ester, 97%
Purity: 97%
CAS No.: 144120-53-6
Molecular Formula: C22H21NO6
Formula Weight: 395.41
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
Beilstein Reference: 5460108
Optical Rotation: -24°(c=1 in DMF)
MDL No.: MFCD00467715
Formula:
C₂₂H₂₁NO₆ Storage Temperature: Ambient |
MDL Number:
MFCD00467715 CAS Number: 144120-53-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...