Boc-DL-Met-Oh 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
N-Boc-DL-methionine
H66372-03
H66372-06
AAH66372-03EA
60.2
USD
AAH66372-03
AAH66372-06
AAH66372-MD
Boc-DL-Met-Oh 98%
Boc-DL-Met-Oh
N-Boc-DL-methionine, 98%
Purity: 98%
CAS No.: 93000-03-4
Molecular Formula: C10H19NO4S
Formula Weight: 249.33
Storage Temperature: 2°C to 8°C
Physical Form: Solid
MDL No.: MFCD00190811
Formula:
C₁₀H₁₉NO₄S MW: 249.325 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00190811 CAS Number: 93000-03-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...