2,3-Butanediol 98%
Supplier: Thermo Scientific Chemicals
Warning
|
(±)-2,3-Butanediol, 98%
Purity: 98%
CAS No.: 513-85-9
Molecular Formula: C4H10O2
Formula Weight: 90.12
Flash Point: 85°(185°F)
Physical Form: Liquid
Density: 1.002
Refractive Index: 1.4330
Sensitivity: Hygroscopic
Appearance: Colorless to pale yellow
Odor: Odorless
Melting Point: 25°
Boiling Point: 183-184°
Beilstein Reference: 969165
MDL No.: MFCD00004523
Formula:
C₄H₁₀O₂ Boiling Pt: 183…184 °C Melting Pt: 25 °C Density: 1.002 Flash Pt: 85 °C(185 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004523 CAS Number: 513-85-9 EINECS: 208-173-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...