Boc-L-β-homovaline 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
N-Boc-L-β-leucine, Boc-β-HoVal-OH, Boc-β-HomoVal-OH, (R)-3-((tert-Butoxycarbonyl)amino)-4-methylpentanoic acid
H66336-03
H66336-MD
AAH66336-03EA
183
USD
AAH66336-03
AAH66336-MD
Boc-L-β-homovaline 95%
Boc-L-beta-homovaline
N-Boc-L-^b-leucine, 95%
Purity: 95%
CAS No.: 183990-64-9
Molecular Formula: C11H21NO4
Formula Weight: 231.29
Physical Form: Solid
Melting Point: 115-120°
Beilstein Reference: 7568192
MDL No.: MFCD01076232
Formula:
C₁₁H₂₁NO₄ MW: 231.292 g/mol Melting Pt: 115…120 °C Storage Temperature: Ambient |
MDL Number:
MFCD01076232 CAS Number: 183990-64-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...