4-Bromonicotinic acid 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Bromopyridine-3-carboxylic acid
H66322-03
H66322-06
AAH66322-03EA
95
USD
AAH66322-03
AAH66322-06
4-Bromonicotinic acid 96%
4-Bromonicotinic acid
4-Bromopyridine-3-carboxylic acid, 96%
Purity: 96%
CAS No.: 15366-62-8
Molecular Formula: C6H4BrNO2
Formula Weight: 202.00
Physical Form: Solid
Appearance: Pale brown to dark brown
Melting Point: 118-134°
MDL No.: MFCD04114236
Formula:
C₆H₄BrNO₂ MW: 202.007 g/mol Melting Pt: 118…134 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD04114236 CAS Number: 15366-62-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...