N-Isopropylacrylamide 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H66262-14
H66262-22
AAH66262-14EA
101.92
USD
AAH66262-14
AAH66262-22
N-Isopropylacrylamide 97%
N-Isopropylacrylamide
N-Isopropylacrylamide, 97%
Purity: 97%
CAS No.: 2210-25-5
Molecular Formula: C6H11NO
Formula Weight: 113.16
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Melting Point: 60-63°
Boiling Point: 89-92°/2mm
Beilstein Reference: 1744926
MDL No.: MFCD00041913
Formula:
C₆H₁₁NO MW: 113.16 g/mol Boiling Pt: 89…92 °C/2 mm Melting Pt: 60…63 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041913 CAS Number: 2210-25-5 EINECS: 218-638-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...