3-Chloro-N-fmoc-L-phenylalanine 95%

Supplier: Thermo Scientific Chemicals

Synonyms: Fmoc-Phe(3-Cl)-OH

H66237-03 H66237-06
AAH66237-03EA 55.3 USD
AAH66237-03 AAH66237-06
3-Chloro-N-fmoc-L-phenylalanine 95%
3-Chloro-N-fmoc-L-phenylalanine

3-Chloro-N-Fmoc-L-phenylalanine, 95%


Purity: 95% CAS No.: 198560-44-0 Molecular Formula: C24H20ClNO4 Formula Weight: 421.88 Storage Temperature: 2°C to 8°C Physical Form: Solid MDL No.: MFCD00672548

Formula: C₂₄H₂₀ClNO₄
MW: 421.877 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00672548
CAS Number: 198560-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR