Fmoc-Pro-Pro-OH
Supplier: Thermo Scientific Chemicals
H66228-03
H66228-06
AAH66228-03EA
94.1
USD
AAH66228-03
AAH66228-06
Fmoc-Pro-Pro-OH
Proteins and Peptides
N-Fmoc-L-prolyl-L-proline, 95%
Purity: 95%
CAS No.: 129223-22-9
Molecular Formula: C25H26N2O5
Formula Weight: 434.49
Storage Temperature: -30°C to -10°C
Physical Form: Solid
MDL No.: MFCD00237673
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...