2-Iodo-5-methoxybenzaldehyde 96%

Supplier: Thermo Scientific Chemicals
Warning

H66163-03 H66163-06
AAH66163-03EA 64 USD
AAH66163-03 AAH66163-06
2-Iodo-5-methoxybenzaldehyde 96%
2-Iodo-5-methoxybenzaldehyde

2-Iodo-5-methoxybenzaldehyde, 96%


Purity: 96% CAS No.: 77287-58-2 Molecular Formula: C8H7IO2 Formula Weight: 262.04 Physical Form: Solid Sensitivity: Air and Light Sensitive Appearance: White to off-white to yel Melting Point: 110-115° MDL No.: MFCD18905098

Formula: C₈H₇IO₂
MW: 262.046 g/mol
Melting Pt: 110…115 °C
Storage Temperature: Ambient
MDL Number: MFCD18905098
CAS Number: 77287-58-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR