3,5-Dimethylisoxazole-4-boronic acid pinacol ester 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3,5-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)isoxazole, 3,5-dimethyl-4-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2-oxazole
H66042-06
H66042-14
AAH66042-06EA
115
USD
AAH66042-06
AAH66042-14
3,5-Dimethylisoxazole-4-boronic acid pinacol ester 97%
3,5-Dimethylisoxazole-4-boronic acid pinacol ester
3,5-Dimethylisoxazole-4-boronic acid pinacol ester, 97%
Purity: 97%
CAS No.: 832114-00-8
Molecular Formula: C11H18BNO3
Formula Weight: 223.08
Physical Form: Solid
Appearance: White to pale brown
Melting Point: 95-99°
MDL No.: MFCD05863910
Formula:
C₁₁H₁₈BNO₃ Melting Pt: 95…99 °C Storage Temperature: Ambient |
MDL Number:
MFCD05863910 CAS Number: 832114-00-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...