5-tert-Butyl-1-pentafluorophenyl-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-glutamate 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
Fmoc-Glu(OtBu)-OPfp, Fmoc-Glu(OtBu)-Opfp, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-glutamic Acid 5-tert-Butyl 1-Pentafluorophenyl Ester, N-Fmoc-L-glutamic Acid 5-tert-Butyl 1-Pentafluorophenyl Ester, 5-tert-Butyl 1-Pentafluorophenyl N-Fmoc-L-glutamate, N-Fmoc-L-glutamic acid 5-tert-butyl ester pentafluorophenyl ester
N-Fmoc-L-glutamic acid 5-tert-butyl ester pentafluorophenyl ester, 98%
Purity: 98%
CAS No.: 86061-04-3
Molecular Formula: C30H26F5NO6
Formula Weight: 591.53
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Beilstein Reference: 3642680
MDL No.: MFCD00065647
Formula:
C₃₀H₂₆F₅NO₆ Storage Temperature: Ambient |
MDL Number:
MFCD00065647 CAS Number: 86061-04-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...