L-Aspartic acid 4-benzyl ester 1-tert-butyl ester hydrochloride 98%

Supplier: Thermo Scientific Chemicals

Synonyms: H-Asp(OBzl)-OtBu·HCl

H65937-03 H65937-06
AAH65937-03EA 113 USD
AAH65937-03 AAH65937-06 AAH65937-MD
L-Aspartic acid 4-benzyl ester 1-tert-butyl ester hydrochloride 98%
L-Aspartic acid 4-benzyl ester 1-tert-butyl ester hydrochloride

L-Aspartic acid 4-benzyl ester 1-tert-butyl ester hydrochloride, 98%


Purity: 98% CAS No.: 94347-11-2 Molecular Formula: C15H21NO4.HCl Formula Weight: 315.79 Storage Temperature: 2°C to 8°C Physical Form: Solid Appearance: White to off-white Beilstein Reference: 2538326 Optical Rotation: +19°(c=2 in methanol) MDL No.: MFCD00190761

Formula: C₁₅H₂₂ClNO₄
Storage Temperature: Ambient
MDL Number: MFCD00190761
CAS Number: 94347-11-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR