L-Cyclopropylglycine 97%

Supplier: Thermo Scientific Chemicals

Synonyms: 2-Cyclopropyl-L-glycine, (S)-2-Amino-2-cyclopropylacetic acid, H-Cyclopropyl-Gly-OH

H65755-03 H65755-06 H65755-MD
AAH65755-03EA 153 USD
AAH65755-03 AAH65755-06 AAH65755-MD
L-Cyclopropylglycine 97%
L-Cyclopropylglycine

2-Cyclopropyl-L-glycine, 97%


Purity: 97% CAS No.: 49606-99-7 Molecular Formula: C5H9NO2 Formula Weight: 115.13 Storage Temperature: 2°C to 8°C Physical Form: Crystalline Appearance: White to off-white Beilstein Reference: 5728866 Optical Rotation: +101°(c=1 in 1M HCl) MDL No.: MFCD06659116

Formula: C₅H₉NO₂
MW: 115.132 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06659116
CAS Number: 49606-99-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR