L-Lysinamide dihydrochloride 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
L-Lysinamide Hydrochloride, H-Lys-NH₂·2HCl, H-Lys-NH₂·2 HCl, (S)-2,6-Diaminohexanamide dihydrochloride
H65741-03
H65741-MD
AAH65741-03EA
227
USD
AAH65741-03
AAH65741-MD
L-Lysinamide dihydrochloride 95%
L-Lysinamide dihydrochloride
L-Lysinamide dihydrochloride, 95%
Purity: 95%
CAS No.: 51127-08-3
Molecular Formula: C6H15N3O.2HCl
Formula Weight: 218.12
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Appearance: Off-white to white
MDL No.: MFCD00058287
Formula:
C₆H₁₇Cl₂N₃O MW: 218.122 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00058287 CAS Number: 51127-08-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...