4-Dodecylaniline 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Dodecylphenylamine, 1-Amino-4-dodecylbenzene
H65528-03
H65528-06
AAH65528-03EA
56.6
USD
AAH65528-03
AAH65528-06
4-Dodecylaniline 95%
4-Dodecylaniline
4-n-Dodecylaniline, 95%
Purity: 95%
CAS No.: 104-42-7
Molecular Formula: C18H31N
Formula Weight: 261.45
Flash Point: 150°(302°F)
Physical Form: Solid
Melting Point: 35-39°
Boiling Point: 220-221°/15mm
Beilstein Reference: 2646305
UN No.: 3077
MDL No.: MFCD00007919
Formula:
C₁₈H₃₁N MW: 261.45 g/mol Boiling Pt: 220…221 °C/15 mm Melting Pt: 35…39 °C Flash Pt: 150 °C (302 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007919 CAS Number: 104-42-7 EINECS: 203-201-3 UN: 3077 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...