L-Phenylalaninamide hydrochloride 97%
Supplier: Thermo Scientific Chemicals
L-Phenylalaninamide hydrochloride, 97%
Purity: 97%
CAS No.: 65864-22-4
Molecular Formula: C9H12N2O.HCl
Formula Weight: 200.67
Physical Form: Powder
Appearance: White
Melting Point: 231-237°
Beilstein Reference: 3915662
MDL No.: MFCD00039083
Formula:
C₉H₁₂N₂O Melting Pt: 231…237 °C Storage Temperature: Ambient |
MDL Number:
MFCD00039083 CAS Number: 65864-22-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...