D-Leucine methyl ester hydrochloride 98%

Supplier: Thermo Scientific Chemicals

Synonyms: H-D-Leu-OMe.HCl

H65456-06 H65456-14 H65456-22
AAH65456-06EA 78.2 USD
AAH65456-06 AAH65456-14 AAH65456-22
D-Leucine methyl ester hydrochloride 98%
D-Leucine methyl ester hydrochloride

D-Leucine methyl ester hydrochloride, 98%


Purity: 98% CAS No.: 5845-53-4 Molecular Formula: C7H15NO2.HCl Formula Weight: 181.66 Physical Form: Solid Sensitivity: Hygroscopic Appearance: Off-white to white Melting Point: 151° Beilstein Reference: 3655228 Optical Rotation: -14°(c=2 in water) MDL No.: MFCD00066115

Formula: C₇H₁₅NO₂
MW: 181.66 g/mol
Melting Pt: 151 °C
Storage Temperature: Ambient
MDL Number: MFCD00066115
CAS Number: 5845-53-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR