Glycyl-L-tyrosine
Supplier: Thermo Scientific Chemicals
H65388-06
H65388-22
AAH65388-06EA
75.6
USD
AAH65388-06
AAH65388-14
AAH65388-22
Glycyl-L-tyrosine
Proteins and Peptides
N-Glycyl-L-tyrosine, 98%
Purity: 98%
CAS No.: 658-79-7
Molecular Formula: C11H14N2O4
Formula Weight: 238.24
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Appearance: White to pale yellow
Beilstein Reference: 2700715
Optical Rotation: +47°(c=1 in water)
MDL No.: MFCD00008126
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...