3-Chloro-N,N-dimethylpropan-1-amine 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
3-Dimethylaminopropyl chloride
H65371-03
H65371-06
H65371-14
AAH65371-03EA
52
USD
AAH65371-03
AAH65371-06
AAH65371-14
3-Chloro-N,N-dimethylpropan-1-amine 97%
3-Chloro-N,N-dimethylpropan-1-amine
3-Dimethylaminopropyl chloride, 97%
Purity: 97%
CAS No.: 109-54-6
Molecular Formula: C5H12ClN
Formula Weight: 121.61
Storage Temperature: -30°C to -10°C
Physical Form: Liquid
Density: 0.929
Refractive Index: 1.4320
Sensitivity: Air Sensitive
Appearance: Colorless
Melting Point: 168°
Beilstein Reference: 605294
MDL No.: MFCD00044496
Formula:
C5H12ClN MW: 121.61 g/mol Melting Pt: 168 °C Density: 0.929 Storage Temperature: Refrigerator |
MDL Number:
MFCD00044496 CAS Number: 109-54-6 EINECS: 203-679-3 UN: 3263 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...