Z-Gln-Gly-OH 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
N-Benzyloxycarbonyl-L-glutaminylglycine, (S)-2-(5-Amino-2-(((benzyloxy)carbonyl)amino)-5-oxopentanamido)acetic acid
H65088-03
H65088-06
H65088-MD
AAH65088-03EA
177
USD
AAH65088-03
AAH65088-06
AAH65088-MD
Z-Gln-Gly-OH 98%
Z-Gln-Gly-OH
N-Benzyloxycarbonyl-L-glutaminylglycine, 98%
Purity: 98%
CAS No.: 6610-42-0
Molecular Formula: C15H19N3O6
Formula Weight: 337.33
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Beilstein Reference: 2179068
MDL No.: MFCD00055926
Formula:
C₁₅H₁₉N₃O₆ MW: 337.332 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00055926 CAS Number: 6610-42-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...