2-Methyl-5-nitrobenzotrifluoride 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Methyl-4-nitro-2-(trifluoromethyl)benzene
H64924-06
H64924-14
AAH64924-03EA
25.1
USD
AAH64924-03
AAH64924-06
AAH64924-14
2-Methyl-5-nitrobenzotrifluoride 98%
2-Methyl-5-nitrobenzotrifluoride
2-Methyl-5-nitrobenzotrifluoride, 98%
Purity: 98%
CAS No.: 89976-12-5
Molecular Formula: C8H6F3NO2
Formula Weight: 205.14
Physical Form: Solid
Sensitivity: Moisture Sensitive
Appearance: Off-white
Beilstein Reference: 2506013
UN No.: 2811
MDL No.: MFCD01631684
Formula:
C₈H₆F₃NO₂ MW: 205.14 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01631684 CAS Number: 89976-12-5 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...