D-Proline ethyl ester hydrochloride 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
H-D-Pro-OEt hydrochloride, (R)-Ethyl pyrrolidine-2-carboxylate hydrochloride
H64910-06
H64910-14
AAH64910-06EA
43.5
USD
AAH64910-06
AAH64910-14
AAH64910-22
D-Proline ethyl ester hydrochloride 98%
D-Proline ethyl ester hydrochloride
D-Proline ethyl ester hydrochloride, 98%
Purity: 98%
CAS No.: 131477-20-8
Molecular Formula: C7H13NO2.HCl
Formula Weight: 179.64
Storage Temperature: -30°C to -10°C
Physical Form: Liquid
Appearance: Pale yellow
Beilstein Reference: 7170607
MDL No.: MFCD00191003
Formula:
C₇H₁₄ClNO₂ MW: 179.644 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00191003 CAS Number: 131477-20-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...