2,5-Dihydroxyterephthalic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

H64888-06 H64888-14
AAH64888-06EA 154.61 USD
AAH64888-06 AAH64888-14
2,5-Dihydroxyterephthalic acid 97%
2,5-Dihydroxyterephthalic acid

2,5-Dihydroxyterephthalic acid, 97%


Purity: 97% CAS No.: 610-92-4 Molecular Formula: C8H6O6 Formula Weight: 198.13 Physical Form: Solid Appearance: Dark grey to green Melting Point: 300° Beilstein Reference: 2696052 MDL No.: MFCD00132933

Formula: C₈H₆O₆
MW: 198.13 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00132933
CAS Number: 610-92-4
EINECS: 210-239-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR