2-Chloro-4,6-dimethylpyridine ≥98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H64733-06
H64733-14
AAH64733-03EA
31.51
USD
AAH64733-03
AAH64733-06
AAH64733-14
2-Chloro-4,6-dimethylpyridine ≥98%
2-Chloro-4,6-dimethylpyridine
2-Chloro-4,6-dimethylpyridine, 98+%
Purity: 98+%
CAS No.: 30838-93-8
Molecular Formula: C7H8ClN
Formula Weight: 141.60
Beilstein Reference: 108937
UN No.: 2810
MDL No.: MFCD08277279
Formula:
C7H8ClN Storage Temperature: Ambient |
MDL Number:
MFCD08277279 CAS Number: 30838-93-8 UN: 2810 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...