2-Amino-5-fluoro-6-methylpyridine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H64700-03
H64700-06
AAH64700-03EA
60.6
USD
AAH64700-03
AAH64700-06
2-Amino-5-fluoro-6-methylpyridine 98%
2-Amino-5-fluoro-6-methylpyridine
2-Amino-5-fluoro-6-methylpyridine, 98%
Purity: 98%
CAS No.: 110919-71-6
Molecular Formula: C6H7FN2
Formula Weight: 126.13
Physical Form: Solid
Appearance: Off-white
Beilstein Reference: 4376910
UN No.: 2811
MDL No.: MFCD09835298
Formula:
C₆H₇FN₂ MW: 126.13 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09835298 CAS Number: 110919-71-6 UN: 2811 ADR: "6.1",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...