6-Hydroxy-1,2,3,4-tetrahydroisoquinoline 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1,2,3,4-Tetrahydroisoquinolin-6-ol
H64512-03
H64512-MD
AAH64512-03EA
400
USD
AAH64512-03
AAH64512-MD
6-Hydroxy-1,2,3,4-tetrahydroisoquinoline 97%
6-Hydroxy-1,2,3,4-tetrahydroisoquinoline
6-Hydroxy-1,2,3,4-tetrahydroisoquinoline, 97%
Purity: 97%
CAS No.: 14446-24-3
Molecular Formula: C9H11NO
Formula Weight: 149.19
Physical Form: Solid
Melting Point: 192-193°
Beilstein Reference: 127578
MDL No.: MFCD01717047
Formula:
C₉H₁₁NO MW: 149.193 g/mol Melting Pt: 192…193 °C Storage Temperature: Ambient |
MDL Number:
MFCD01717047 CAS Number: 14446-24-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...