2-[2-(Trifluoromethyl)phenyl]ethylamine 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H64492-03
H64492-06
H64492-14
AAH64492-03EA
53.2
USD
AAH64492-03
AAH64492-06
AAH64492-14
2-[2-(Trifluoromethyl)phenyl]ethylamine 95%
2-[2-(Trifluoromethyl)phenyl]ethylamine
2-[2-(Trifluoromethyl)phenyl]ethylamine, 95%
Purity: 95%
CAS No.: 774-18-5
Molecular Formula: C9H10F3N
Formula Weight: 189.18
Physical Form: Liquid
Appearance: Pale yellow
Beilstein Reference: 7202211
UN No.: 2735
MDL No.: MFCD00128232
Formula:
C₉H₁₀F₃N Storage Temperature: Ambient |
MDL Number:
MFCD00128232 CAS Number: 774-18-5 EINECS: 212-262-5 UN: 2735 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...