4-iodobenzene-1,2-diamine 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Iodo-o-phenylenediamine, 4-Iodobenzene-1,2-diamine
H64469-03
H64469-06
AAH64469-03EA
117
USD
AAH64469-03
AAH64469-06
AAH64469-MD
4-iodobenzene-1,2-diamine 95%
4-iodobenzene-1,2-diamine
4-Iodo-o-phenylenediamine, 95%
Purity: 95%
CAS No.: 21304-38-1
Molecular Formula: C6H7IN2
Formula Weight: 234.04
Physical Form: Solid
Sensitivity: Light Sensitive
Melting Point: 73-77°
Beilstein Reference: 2716284
UN No.: 2811
MDL No.: MFCD08669466
Formula:
C₆H₇IN₂ MW: 234.04 g/mol Melting Pt: 73…77 °C Storage Temperature: Ambient |
MDL Number:
MFCD08669466 CAS Number: 21304-38-1 UN: 2811 ADR: "6.1",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...