2-(3-Trifluoromethylphenyl)ethylamine 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
3-(Trifluoromethyl)phenethylamine
H64084-03
H64084-06
H64084-14
AAH64084-03EA
37.2
USD
AAH64084-03
AAH64084-06
AAH64084-14
2-(3-Trifluoromethylphenyl)ethylamine 98%
2-(3-Trifluoromethylphenyl)ethylamine
2-[3-(Trifluoromethyl)phenyl]ethylamine, 98%
Purity: 98%
CAS No.: 52516-30-0
Molecular Formula: C9H10F3N
Formula Weight: 189.18
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Density: 1.18
Refractive Index: 1.4645
Sensitivity: Light Sensitive
Appearance: Colorless to pale yellow
Boiling Point: 113°/22mm
Beilstein Reference: 4398117
UN No.: 2735
MDL No.: MFCD00040756
Formula:
C₉H₁₀F₃N Boiling Pt: 113 °C/22 mm Density: 1.18 Storage Temperature: Ambient |
MDL Number:
MFCD00040756 CAS Number: 52516-30-0 EINECS: 257-988-3 UN: 2735 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...