2,5-Dichloropyrimidine 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H64066-06
H64066-14
H64066-22
AAH64066-06EA
42.5
USD
AAH64066-06
AAH64066-14
AAH64066-22
2,5-Dichloropyrimidine 95%
2,5-Dichloropyrimidine
2,5-Dichloropyrimidine, 95%
Purity: 95%
CAS No.: 22536-67-0
Molecular Formula: C4H2Cl2N2
Formula Weight: 148.97
Flash Point: 107°(225°F)
Physical Form: Powder
Appearance: Off-white
Melting Point: 48-55°
Beilstein Reference: 110912
UN No.: 3077
MDL No.: MFCD00673131
Formula:
C4H2Cl2N2 MW: 148.97 g/mol Melting Pt: 48…55 °C Flash Pt: 107 °C(225 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00673131 CAS Number: 22536-67-0 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...