(1-Indolyl)acetic acid 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H63935-03
H63935-MD
AAH63935-03EA
217
USD
AAH63935-03
AAH63935-MD
(1-Indolyl)acetic acid 95%
(1-Indolyl)acetic acid
1-Indoleacetic acid, 95%
Purity: 95%
CAS No.: 24297-59-4
Molecular Formula: C10H9NO2
Formula Weight: 175.18
Physical Form: Solid
Beilstein Reference: 142924
MDL No.: MFCD00047262
Formula:
C₁₀H₉NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD00047262 CAS Number: 24297-59-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...