3,4-Dichloropyridine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H63749-03
H63749-06
AAH63749-03EA
36.8
USD
AAH63749-03
AAH63749-06
3,4-Dichloropyridine 98%
3,4-Dichloropyridine
3,4-Dichloropyridine, 98%
Purity: 98%
CAS No.: 55934-00-4
Molecular Formula: C5H3Cl2N
Formula Weight: 147.99
Physical Form: Liquid
Appearance: Colorless
UN No.: 2810
MDL No.: MFCD01861989
Formula:
C₅H₃Cl₂N Boiling Pt: 182…183 °C Melting Pt: 22…24 °C Storage Temperature: Ambient |
MDL Number:
MFCD01861989 CAS Number: 55934-00-4 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...