1-Acetyl-5-bromoindole 97%
Supplier: Thermo Scientific Chemicals
H63745-03
H63745-MD
AAH63745-03EA
122
USD
AAH63745-03
AAH63745-MD
1-Acetyl-5-bromoindole 97%
1-Acetyl-5-bromoindole
1-Acetyl-5-bromoindole, 97%
Purity: 97%
CAS No.: 61995-52-6
Molecular Formula: C10H8BrNO
Formula Weight: 238.08
Physical Form: Solid
Beilstein Reference: 1283925
MDL No.: MFCD00238530
Formula:
C₁₀H₈BrNO Storage Temperature: Ambient |
MDL Number:
MFCD00238530 CAS Number: 61995-52-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...