3-aminopyrazine-2-carboxamide 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Aminopyrazine-2-carboxamide

H63553-03 H63553-06
AAH63553-03EA 191 USD
AAH63553-03 AAH63553-06 AAH63553-MD
3-aminopyrazine-2-carboxamide 96%
3-aminopyrazine-2-carboxamide

3-Aminopyrazine-2-carboxamide, 96%


Purity: 96% CAS No.: 32587-10-3 Molecular Formula: C5H6N4O Formula Weight: 138.13 Physical Form: Solid MDL No.: MFCD03407446

Formula: C₅H₆N₄O
Storage Temperature: Ambient
MDL Number: MFCD03407446
CAS Number: 32587-10-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR