4-(Bromomethyl)-3-fluorobenzonitrile 95%

Supplier: Thermo Scientific Chemicals
Danger

H63245-03 H63245-06
AAH63245-03EA 35.6 USD
AAH63245-03 AAH63245-06
4-(Bromomethyl)-3-fluorobenzonitrile 95%
4-(Bromomethyl)-3-fluorobenzonitrile

4-Bromomethyl-3-fluorobenzonitrile, 95%


Purity: 95% CAS No.: 105942-09-4 Molecular Formula: C8H5BrFN Formula Weight: 214.04 Storage Temperature: -30°C to -10°C Physical Form: Solid Appearance: Off-white Melting Point: 75-79° dec Beilstein Reference: 7758134 UN No.: 2923 MDL No.: MFCD07368341

Formula: C₈H₅BrFN
MW: 214.04 g/mol
Melting Pt: 75…79 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD07368341
CAS Number: 105942-09-4
UN: 2923
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR