5-bromoquinolin-6-amine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
6-Amino-5-bromoquinoline, 5-Bromoquinolin-6-amine
H63150-03
H63150-06
AAH63150-03EA
185
USD
AAH63150-03
AAH63150-06
AAH63150-MD
5-bromoquinolin-6-amine 97%
5-bromoquinolin-6-amine
6-Amino-5-bromoquinoline, 97%
Purity: 97%
CAS No.: 50358-42-4
Molecular Formula: C9H7BrN2
Formula Weight: 223.07
Physical Form: Solid
Appearance: Yellow to brown
MDL No.: MFCD00234503
Formula:
C₉H₇BrN₂ MW: 223.073 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00234503 CAS Number: 50358-42-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...