3-(1-Methyl-4-piperidinyl)indole 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
3-(1-Methyl-4-piperidinyl)indole, 97%
Purity: 97%
CAS No.: 17403-07-5
Molecular Formula: C14H18N2
Formula Weight: 214.31
Physical Form: Solid
Appearance: Off-white to yellow
Beilstein Reference: 1573327
MDL No.: MFCD07776765
Formula:
C₁₄H₁₈N₂ Storage Temperature: Ambient |
MDL Number:
MFCD07776765 CAS Number: 17403-07-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...