2,3,4,5-Tetrafluoroaniline 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H61417-03
H61417-06
AAH61417-03EA
66.8
USD
AAH61417-03
AAH61417-06
2,3,4,5-Tetrafluoroaniline 98%
2,3,4,5-Tetrafluoroaniline
2,3,4,5-Tetrafluoroaniline, 98%
Purity: 98%
CAS No.: 5580-80-3
Molecular Formula: C6H3F4N
Formula Weight: 165.09
Flash Point: 79°(174°F)
Physical Form: Powder
Melting Point: 27-29°
Boiling Point: 78°
UN No.: 2811
MDL No.: MFCD00025153
Formula:
C₆H₃F₄N MW: 165.09 g/mol Boiling Pt: 78 °C Melting Pt: 27…29 °C Flash Pt: 79 °C (174 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00025153 CAS Number: 5580-80-3 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...