Bis[2-(di-t-butylphosphino)ethyl]amine 10% (w/w) in THF

Supplier: Thermo Scientific Chemicals
Danger

H60493-03 H60493-06 H60493-14
AAH60493-03EA 44.3 USD
AAH60493-03 AAH60493-06 AAH60493-14
Bis[2-(di-t-butylphosphino)ethyl]amine 10% (w/w) in THF
Bis[2-(di-t-butylphosphino)ethyl]amine

Bis[2-(di-tert-butylphosphino)ethyl]amine, 10% w/w soln. in THF


  • Refractive Index: 1.4180
  • Odour: Unpleasant


CAS No.: 944710-34-3 Molecular Formula: C20H45NP2 Formula Weight: 361.53 Refractive Index: 1.4180 Odor: Unpleasant UN No.: 1993 MDL No.: MFCD17014018

Formula: C₂₀H₄₅NP₂
MW: 361.535 g/mol
Density: 0.911
Storage Temperature: Ambient
MDL Number: MFCD17014018
CAS Number: 944710-34-3
UN: 1993
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR