5-Chlorovaleryl chloride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
5-Chlorovaleroyl chloride, 98%
Purity: 98%
CAS No.: 1575-61-7
Molecular Formula: C5H8Cl2O
Formula Weight: 155.02
Flash Point: 91°(196°F)
Refractive Index: 1.464
Boiling Point: 38-39°
Beilstein Reference: 1745182
UN No.: 3265
MDL No.: MFCD00000758
Formula:
C₅H₈Cl₂O Boiling Pt: 38…39 °C Density: 1.21 g/cm³ (20 °C) Flash Pt: 91 °C (196 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000758 CAS Number: 1575-61-7 EINECS: 216-403-1 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...