4-Fluoro-N-methylaniline 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N-(4-Fluorophenyl)-N-methylamine

H55105-03 H55105-06
AAH55105-03EA 41.2 USD
AAH55105-03 AAH55105-06 AAH55105-14
4-Fluoro-N-methylaniline 97%
4-Fluoro-N-methylaniline

4-Fluoro-N-methylaniline, 97%


Purity: 97% CAS No.: 459-59-6 Molecular Formula: C7FH8N Formula Weight: 125.15 Flash Point: 76°(169°F) Refractive Index: 1.5320 Boiling Point: 79°/11mm UN No.: 2810 MDL No.: MFCD00017942

Formula: C₇H₈FN
MW: 125.15 g/mol
Boiling Pt: 79 °C/11 mm
Density: 1.040
Flash Pt: 76 °C (169 °F)
Storage Temperature: Ambient
MDL Number: MFCD00017942
CAS Number: 459-59-6
EINECS: 207-294-1
UN: 2810
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR