1,3-Di-tert-butyl-2-methoxybenzene 99%
Supplier: Thermo Scientific Chemicals
H37487-09
H37487-18
H37487-30
AAH37487-09EA
46.4
USD
AAH37487-09
AAH37487-18
AAH37487-30
1,3-Di-tert-butyl-2-methoxybenzene 99%
1,3-Di-tert-butyl-2-methoxybenzene
1,3-Di-tert-butyl-2-methoxybenzene, 99%
Purity: 99%
CAS No.: 1516-95-6
Molecular Formula: C15H24O
Formula Weight: 220.35
Physical Form: Liquid
Appearance: Colorless to pale yellow
Boiling Point: 124°/9mm
MDL No.: MFCD20483412
Formula:
C₁₅H₂₄O Boiling Pt: 124 °C/9 mm Storage Temperature: Ambient |
MDL Number:
MFCD20483412 CAS Number: 1516-95-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...