(R)-(+)-5,5',6,6',7,7',8,8'-Octahydro-1,1'-bi-2-naphthol ≥98%, ee 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(R)-(+)-2,2'-Dihydroxy-5,5',6,6',7,7',8,8'-octahydro-1,1'-binaphthyl, (R)-5,5',6,6',7,7',8,8'-Octahydro[1,1'-binaphthalene]-2,2'-diol
H36142-03
H36142-06
AAH36142-03EA
112
USD
AAH36142-03
AAH36142-06
(R)-(+)-5,5',6,6',7,7',8,8'-Octahydro-1,1'-bi-2-naphthol ≥98%, ee 99%
(R)-(+)-5,5',6,6',7,7',8,8'-Octahydro-1,1'-bi-2-naphthol
(R)-(+)-5,5',6,6',7,7',8,8'-Octahydro-1,1'-bi-2-naphthol, 98%, 99+% ee
Purity: 98%,
CAS No.: 65355-14-8
Molecular Formula: C20H22O2
Formula Weight: 294.39
Physical Form: Solid
Sensitivity: Hygroscopic
Appearance: White
Melting Point: 160-166°
MDL No.: MFCD02093485
Formula:
C₂₀H₂₂O₂ MW: 294.39 g/mol Melting Pt: 160…166 °C Storage Temperature: Ambient |
MDL Number:
MFCD02093485 CAS Number: 65355-14-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...